Anti-CCL18 Rabbit Monoclonal Antibody [clone: EPR24781-84]

Supplier: Abcam
AB300057-100 AB300057-20UL AB300057-1ML
ABCAAB300057-100EA 707 EUR
ABCAAB300057-100 ABCAAB300057-20UL ABCAAB300057-1ML
Anti-CCL18 Rabbit Monoclonal Antibody [clone: EPR24781-84]
Antibodies
Rabbit monoclonal [EPR24781-84] to CCL18.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CCL18
Clonality: Monoclonal
Clone: EPR24781-84
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR