Anti-SIAH1 Rabbit Monoclonal Antibody [clone: EPR27101-45]

Supplier: Abcam
AB305249-10 AB305249-100 AB305249-20UL AB305249-1ML
ABCAAB305249-10EA 219 EUR
ABCAAB305249-10 ABCAAB305249-100 ABCAAB305249-20UL ABCAAB305249-1ML
Anti-SIAH1 Rabbit Monoclonal Antibody [clone: EPR27101-45]
Antibodies
Rabbit monoclonal [EPR27101-45] to SIAH1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: SIAH1
Clonality: Monoclonal
Clone: EPR27101-45
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR