Anti-CD272/BTLA Rabbit Monoclonal Antibody [clone: EPR25380-391]

Supplier: Abcam
AB303540-100 AB303540-10 AB303540-20UL AB303540-1ML
ABCAAB303540-100EA 707 EUR
ABCAAB303540-100 ABCAAB303540-10 ABCAAB303540-20UL ABCAAB303540-1ML
Anti-CD272/BTLA Rabbit Monoclonal Antibody [clone: EPR25380-391]
Antibodies
Rabbit monoclonal [EPR25380-391] to CD272/BTLA.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD272/BTLA
Clonality: Monoclonal
Clone: EPR25380-391
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR