Anti-Laminin beta 1 Rabbit Monoclonal Antibody [clone: EPR3189(2)]

Supplier: Abcam
AB247686-100 AB247686-1
ABCAAB247686-100EA 1020 EUR
ABCAAB247686-100 ABCAAB247686-1
Anti-Laminin beta 1 Rabbit Monoclonal Antibody [clone: EPR3189(2)]
Antibodies
Rabbit monoclonal [EPR3189(2)] to Laminin beta 1 - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Laminin beta 1
Clonality: Monoclonal
Clone: EPR3189(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR