Anti-Human Papillomavirus 16 (E7) Rabbit Monoclonal Antibody [clone: EPR25814-25]

Supplier: Abcam
AB300763-100 AB300763-1
ABCAAB300763-100EA 900 EUR
ABCAAB300763-100 ABCAAB300763-1
Anti-Human Papillomavirus 16 (E7) Rabbit Monoclonal Antibody [clone: EPR25814-25]
Antibodies
Rabbit monoclonal [EPR25814-25] to Human Papillomavirus 16 (E7) - BSA and Azide free (Detector).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Human Papillomavirus 16 (E7)
Clonality: Monoclonal
Clone: EPR25814-25
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: HPV16
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR