Anti-POLE Rabbit Monoclonal Antibody [clone: EPR28639-64]

Supplier: Abcam
AB318969-100UG AB318969-1MG
ABCAAB318969-100UGEA 1020 EUR
ABCAAB318969-100UG ABCAAB318969-1MG
Anti-POLE Rabbit Monoclonal Antibody [clone: EPR28639-64]
Antibodies
Rabbit monoclonal [EPR28639-64] to POLE - BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: POLE
Clonality: Monoclonal
Clone: EPR28639-64
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR