Anti-ECM Isoform 1 Rabbit Monoclonal Antibody [clone: EPR25810-46]

Supplier: Abcam
AB314893-20UL AB314893-100UL AB314893-1ML
ABCAAB314893-20ULEA 244 EUR
ABCAAB314893-20UL ABCAAB314893-100UL ABCAAB314893-1ML
Anti-ECM Isoform 1 Rabbit Monoclonal Antibody [clone: EPR25810-46]
Antibodies
Rabbit monoclonal [EPR25810-46] to ECM Isoform 1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ECM Isoform 1
Clonality: Monoclonal
Clone: EPR25810-46
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR