Anti-AIRE Rabbit Monoclonal Antibody [clone: EPR28045-139]

Supplier: Abcam
AB315165-1MG AB315165-100UG
ABCAAB315165-1MGEA 9160 EUR
ABCAAB315165-1MG ABCAAB315165-100UG
Anti-AIRE Rabbit Monoclonal Antibody [clone: EPR28045-139]
Rabbit monoclonal [EPR28045-139] to AIRE - BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: AIRE
Clonality: Monoclonal
Clone: EPR28045-139
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR