Anti-Carbonic Anhydrase 8/CA8 Rabbit Monoclonal Antibody [clone: EPR29175-67]

Supplier: Abcam
AB319156-20UL AB319156-100UL AB319156-1ML
ABCAAB319156-20ULEA 244 EUR
ABCAAB319156-20UL ABCAAB319156-100UL ABCAAB319156-1ML
Anti-Carbonic Anhydrase 8/CA8 Rabbit Monoclonal Antibody [clone: EPR29175-67]
Antibodies
Rabbit monoclonal [EPR29175-67] to Carbonic Anhydrase 8/CA8.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Carbonic Anhydrase 8/CA8
Clonality: Monoclonal
Clone: EPR29175-67
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR