Anti-p53 (acetyl K305 + K370 + K373) Rabbit Recombinant Multiclonal Antibody [clone: RM1200]

Supplier: Abcam
AB321819-20UL AB321819-100UL AB321819-1ML
ABCAAB321819-20ULEA 207 EUR
ABCAAB321819-20UL ABCAAB321819-100UL ABCAAB321819-1ML
Anti-p53 (acetyl K305 + K370 + K373) Rabbit Recombinant Multiclonal Antibody [clone: RM1200]
Antibodies
Rabbit recombinant multiclonal [RM1200] to p53 (acetyl K305 + K370 + K373).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: p53
Clonality: Recombinant Multiclonal
Clone: RM1200
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR