Anti-Transmembrane 4 L6 family member 1 Rabbit Monoclonal Antibody [clone: EPR28894-539]

Supplier: Abcam
AB322342-20UL AB322342-100UL AB322342-1ML
ABCAAB322342-20ULEA 244 EUR
ABCAAB322342-20UL ABCAAB322342-100UL ABCAAB322342-1ML
Anti-Transmembrane 4 L6 family member 1 Rabbit Monoclonal Antibody [clone: EPR28894-539]
Antibodies
Rabbit monoclonal [EPR28894-539] to Transmembrane 4 L6 family member 1.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Transmembrane 4 L6 family member 1
Clonality: Monoclonal
Clone: EPR28894-539
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR