Anti-p75 NGF Receptor Rabbit Recombinant Multiclonal Antibody [clone: EPR26732-M2]

Supplier: Abcam
AB320860-100UG AB320860-1MG
ABCAAB320860-100UGEA 919 EUR
ABCAAB320860-100UG ABCAAB320860-1MG
Anti-p75 NGF Receptor Rabbit Recombinant Multiclonal Antibody [clone: EPR26732-M2]
Antibodies
Rabbit recombinant multiclonal [EPR26732-M2] to p75 NGF Receptor - BSA and Azide free (Detector).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: p75 NGF Receptor
Clonality: Recombinant Multiclonal
Clone: EPR26732-M2
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR