Anti-FTL Rabbit Monoclonal Antibody [clone: EPR26117-110] (Alexa Fluor® 647)

Supplier: Abcam
AB318240-10UL AB318240-100UL
ABCAAB318240-10ULEA 238 EUR
ABCAAB318240-10UL ABCAAB318240-100UL
Anti-FTL Rabbit Monoclonal Antibody [clone: EPR26117-110] (Alexa Fluor® 647)
Antibodies
Alexa Fluor® 647 Rabbit monoclonal [EPR26117-110] to Ferritin Light Chain.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: FTL
Clonality: Monoclonal
Clone: EPR26117-110
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR