Anti-beta Catenin (phospho S675) Rabbit Monoclonal Antibody [clone: EPR28410-45]

Supplier: Abcam
AB314450-20UL AB314450-100UL AB314450-1ML
ABCAAB314450-20ULEA 169 EUR
ABCAAB314450-20UL ABCAAB314450-100UL ABCAAB314450-1ML
Anti-beta Catenin (phospho S675) Rabbit Monoclonal Antibody [clone: EPR28410-45]
Antibodies
Rabbit monoclonal [EPR28410-45] to beta Catenin (phospho S675).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: beta Catenin
Clonality: Monoclonal
Clone: EPR28410-45
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR