Anti-p53 (acetyl K305 + K370 + K373) Rabbit Recombinant Multiclonal Antibody [clone: RM1200]

Supplier: Abcam
AB321820-100UG AB321820-1MG
ABCAAB321820-100UGEA 1020 EUR
ABCAAB321820-100UG ABCAAB321820-1MG
Anti-p53 (acetyl K305 + K370 + K373) Rabbit Recombinant Multiclonal Antibody [clone: RM1200]
Antibodies
Rabbit recombinant multiclonal [RM1200] to p53 (acetyl K305 + K370 + K373) - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: p53
Clonality: Recombinant Multiclonal
Clone: RM1200
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR