TMB ELISA Substrate (Fast Kinetic Rate)
Supplier: Abcam
AB171524-100 AB171524-1000
ABCAAB171524-100EA 100 EUR
ABCAAB171524-100 ABCAAB171524-1000
TMB ELISA Substrate (Fast Kinetic Rate)
Assays ELISAs
Abcam's TMB ELISA Substrate (Fast Kinetic Rate) detects horseradish peroxidase (HRP) activity and contains 3,3',5,5'-tetramethylbenzidine in a mildly acidic buffer.