Density standards
182 density standards for use at different temperatures (15°C, 20°C, 25°C, 40°C, 50°C, 60°, 80°C)
Extensive range of density standards in accordance with ASTM D1480-12 for density or relative density (specific gravity) measurements by Bingham Pycnometer.
- High accuracy
- Verified using Bingham Pycnometer
- All products tested in accordance with ASTM D1480-12 guidelines
- Consistent, traceable, certified
- Certificates of Analysis and Safety Data Sheets available online