Minimate™ Tangential Flow Filtration System
Minimate™ Tangential Flow Filtration System Streamline laboratory-scale concentration, desalting, and buffer exchange processes.
picture from research brochure page 10
- System’s plug-n-play design includes all the hardware, tubing, and fittings needed to get your TFF process up and running quickly
- Concentration and diafiltration processes can be performed on the same system with minimal user intervention
- Cost-effective design − easy to clean and reuse
Applications
- Concentrate and desalt proteins, peptides, or nucleic acids (DNA, RNA, oligonucleotides)
- Recover antibodies or recombinant proteins from clarified cell culture media
- Separate (fractionate) large from small biomolecules
- Concentrate viruses or gene therapy vectors
- Prepare samples prior to column chromatography