Extensive refractive index standard range from (1,34325-1,65808 ŋD)
A complete range of 25 standards solvent-based and sucrose-stabilised at 20°C
These 25 refractive index standards can be used with any brand of refractometer.
- One of the most extensive range in the marketplace
- Presented in a convenient high quality 15 ml dropper bottle
- Uncertainty of measurement up to ±0,00014 ŋD units
- Consistency of product: Independent, traceable, certified
- Certificates of Analysis and Safety Data Sheets available online