Quantus Underwear Standard
Features Sweatshirt:
  • Lap seams for tight and secure seam-enclosure
  • Antistatic
  • Long sleeves
  • Also available as T-Shirt with short sleeves
  • Knitted cuffs
  • Round neck, knitted
  • Extra material flap for bar code inside
  • Antimicrobial finish
  • Color: dark blue
  • Fabric: Microfibre (100% Polyester with carbon)
Sizes: XS - XXXL
Features Trousers:
  • Antistatic
  • Wide, double-stitched comfort waist band
  • Rubber band at ankle height
  • Extra material insert/pocket positioned under the waistband on the right (for bar code)
  • Antimicrobial finish
Color: dark blue
Fabrics: Microfibre (100% Polyester with carbon)
Sizes: XS - XXXL
Contact us