Double gloving
INCREASED PROTECTION BY DOUBLE GLOVING
In order to increase the level of protection while handling cytotoxic material, a double-gloving system is recommended. Glove defects are often not identified at the time of incident. Wearing gloves of two different colours allows the operator to immediately identify a breach and change gloves.
Pictures 1 to 5 show the single steps on how to put on the second layer of gloves. For the choice of the product itself, there is not a "best practice". The operator may need to try several different combinations of gloves and sizes. i.e. some operators may find that a larger glove inside and a smaller outside works well for them, others may choose the same size for both inside and outside.
For further consultation on the best choice of glove and the gloving procedure, please contact us at cleanroom@eu.vwr.com
OUR RECOMMENDATIONS:
WHICH CLEANROOM GLOVES TO USE IN COMBINATION FOR DOUBLE GLOVING
![]() |
Cleanroom gloves, TouchNTuff® 93-700 |
|
![]() |
Cleanroom gloves, TouchNTuff® 93-700 |
![]() |
Cleanroom gloves, BioClean Ultimate™ |
|
![]() |
Cleanroom gloves, BioClean Fusion™ |
![]() |
Cleanroom gloves, DermaShield® 73-711 / 73-721 |
|
![]() |
Gloves, TouchNTuff® 73-300/73-500 |
![]() |
Cleanroom gloves, KIMTECH PURE* Sterile G3 |
|
![]() |
Cleanroom gloves, KIMTECH PURE* G3 Sterile STERLING* |