Total Organic Carbon (TOC)
Total Organic Carbon (TOC) is the amount of carbon bound in an organic compound and is often used as a non-specific indicator of water quality. The rapid method of TOC determination by colorimetric method makes this an easy method to ensure great results.
- Rapid digestion
- Choice of photometers and spectrophotometers and ready-to-use TOC tubes and powder pillows.
Thermoreactors
Rapid digestion for COD and TOC...
Photometers & Spectrophotometers
VWR offers a large choice of systems to measure the different elements of water...
Merck TOC Tubes
User friendly with virtually all of the reagents necessary for the analysis contained in cells...
Solutions For TOC Analysis
Total Organic Carbon (TOC) analysis solutions for detecting organic contamination in water...