Leica Compound Upright Microscopes
Get insights into the smallest details and inspect and document results efficiently with Life Science  compound microscopes from Leica Microsystems. Each solution can be customised with brilliant, cool LED illumination, ergonomic accessories, sophisticated digital cameras and intuitive software to meet a broad range of applications.
Request a call
DM300 upright microscope
Monocular, binocular or trinocular educational microscope for life sciences with four achromat or plan achromat objectives in life science classrooms.
More info
DM500 upright microscope
Binocular or trinocular educational microscope for life science students with four 'infinity corrected' plan-achromat objectives and fluorescence capability.
More info
DM750 upright microscope
Binocular or trinocular educational microscope for life science with four or five 'infinity corrected' or HI plan-achromat objectives and fluorescence capability.
More info
DM1000 LED upright microscope
The ergonomic DM1000 LED laboratory microscope is perfect for clinical laboratory applications such as histology, cytology, haematology and pathology.
More info
DM2000 LED upright microscopes
With their modular design and high performance fluorescence, Leica DM2000 LED microscopes are ideal for complex tasks in pathology, cytology and much more.
More info