Automation Tips for Liquid Handling Systems
With hundreds of Thermo Scientific™ automation tips designed to fit over 50 workstations and pipetting head variations, we're certain you will find tips that increase efficiency through accurate and precise performance.
These tips undergo a rigorous quality control program to ensure consistency resulting in superior tip straightness and a low coefficient of variation ranging from 1% to 3%. With stringent manufacturing specifications, our tips adhere to tolerances 400% tighter than standard pipette tips.
Thermo Scientific Automation Tips for Beckman Automated Liquid Handling Platforms
- Tips are validated on the actual workstations for which they are created
- Available in sterile, nonsterile, sterile filtered, wide bore, and low retention
- Extended tip length is compatible with 96-well and 384-well deep well plates
Thermo Scientific Automation Tips for Tecan Automated Liquid Handling Platforms
- Conductive option provides increased pipetting performance
- Tips are validated on the actual workstations for which they are created
- 20 μl tips increase precision of low volume pipetting through minimization of dead air space
Thermo Scientific Automation Tips for PerkinElmer/Packard Robotic Automated Liquid Handling Platforms
- Conductive option provides increased pipetting performance
- Increased precision for your toughest multi-dispensing assays
- Tips are validated on the actual workstations for which they are created
Thermo Scientific Automation Tips for Agilent Automated Liquid Handling Platforms
- Low percentage coefficient of variation
- Available in nonsterile, sterile or sterile filtered
- Validated on the actual workstation for which the tip is created
Thermo Scientific Automation Tips for Eppendorf Automated Liquid Handling Platforms
- Low percentage coefficient of variation
- Available in nonsterile, sterile or sterile filtered
- Validated on the actual workstation for which the tip is created
Thermo Scientific Automation Tips for other Automated Liquid Handling Platforms
Tips for:
- Qiagen Universal Systems
- Molecular Devices Systems
- Caliper/Zymark Liquid Handling Workstations
- BioTek Pipetting Systems
- Thermo Scientific Automated Liquid Handling Platforms