Sampling accessories
The right accessories will help you to round-off your sampling process. This chapter shows a small selection of useful tools to optimise your sampling.
Close-it labels
Stick firmly to most materials, even to surfaces that are slightly moist or coated with powder. Ideal for QC and repairing damaged packaging.
Seals
Useable as original seals and protection from sample manipulation. The initial opening of the packaging is made obvious and irrevocably visible.
Sampling bags and tubes
It is not only samplers that are crucial in the sampling process, sample transport containers are essential for sample quality.
Cleaning brushes
Help to clean different kind of sampling lances thoroughly. Equipped with strong efficient bristles.
Snowpack dry ice machine
Enables you to obtain a -79 °C dry ice tablet. Ideal for lowering temperatures within cooling baths or boxes for sample storage or transport.
Accessories for zone samplers
These special accessories help to collect and release samples.