Safety eyewear
Overspecs, full vision goggles and safety spectacles.
Modern eye protection bringing together safety, functionality, quality and design, in order to keep pace with the rapid advances in industry and construction – this is what our customers expect of us, and what we expect of ourselves.
Sportstyle spectacles
Sports-style safety spectacles with anti-scratch, anti-fog and anti-uv lens.
I works spectacles
100% metal-free, low weight safety spectacles with panoramic lens.
Pheos CX2 spectacles
Comfortable, modern safety spectacles with twist-temple technology for a perfect and non-slip fit.
Ultrasonic CR goggles
Autoclavable cleanroom goggles with indirect ventilation.
super f OTG CR
Autoclavable and ultra-light overspecs with panoramic lens
pheos cx2 sonic
lightest compact wide-vision goggles with very soft textile headband for secure and comfortable fit