Myco-Sniff™ Mycoplasma PCR Kit, MP Biomedicals
Supplier: MP Biomedicals
093050201
093050201.EA 494 EUR
093050201.
Myco-Sniff™ Mycoplasma PCR Kit, MP Biomedicals
Assays qPCR Assays
Myco-Sniff™ mycoplasma PCR detection kit provides a highly sensitive, specific and rapid method for the detection of a broad range of mycoplasma species in cell cultures.
  • Highly specific: No Interference of animal or bacterial DNA
  • Sample control: Easily verify the effectiveness of template gDNA by checking the amplification of the sample control DNA
The detection limit for contaminating mycoplasma is as low as 20 CFU/ml. Includes 48 lyophilised PCR tubes for 20 μl reactions.
Order Now
SPECIFICATIONS
Environmentally Preferable