Stoppers, press down, VacuCap®
Supplier: Simport Scientific
T402-13N T402-13R T402-13B T402-13G T402-13Y T402-13GY T402-16N T402-16R T402-16B T402-16G T402-16Y T402-16L T402-16GY
217-0101EA 40.5 EUR
217-0101 720-0552 217-0098 217-0099 217-0102 720-0551 720-0554 720-0555 217-0103 217-0104 217-0106 217-0105 720-0553
Stoppers, press down, VacuCap®
Stoppers PE/LDPE/HDPE Stoppers
Ideal closures for glass and plastic reagent, sample and culture tubes, LDPE.
  • Disposable seals are an inexpensive alternative to standard seals
  • Double flanged seal for an impermeable, secure fit
  • Colour coded for sample identification
Flexible to protect from aerosols or highly infectious microorganisms. Samples are protected from cross-contamination and the work environment from infection due to leaks. The double flanged seal guarantees a tight and impermeable fit for a wide range of applications, including centrifuging.
Order Now
SPECIFICATIONS
Environmentally Preferable