Anti-IgG Sheep Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Tonbo Biosciences
35-8078-M002
35-8078-M002EA 195 EUR
35-8078-M002
Anti-IgG Sheep Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Sheep anti-Rabbit IgG (H&L) - Affinity Pure, FITC Conjugate

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Heavy and light chain
Host: Sheep
Isotype:
Reactivity: Rabbit
Order Now

SPECIFICATIONS

Antigen symbol IgG
Environmentally Preferable
Antigen name Immunoglobulin G
Antibody type Secondary
Clonality Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Rabbit
Host Sheep
Isotype
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Epitope Heavy and light chain
Storage temperature Store at 2-8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR