VWR® PerfectBlue Twin M, Dual-Gel System

Supplier: VWR Collection
700-0955EA 2300 EUR
700-0955 700-0961
VWR® PerfectBlue Twin M, Dual-Gel System
Electrophoresis Systems Vertical Gel Electrophoresis Systems
For 16×14 cm gels, the Twin M is a versatile system for SDS PAGE applications, in addition to non denaturing electrophoresis of proteins, SSCPs and dinucleotide repeats. With the enlarged upper buffer chamber, water circulation system and heat-sinking aluminium ceramic plates, the Twin M provides high speed separation while maintaining straight, sharp bands.

*Based on an average gel and a tooth at 20 mm depth

Ordering information: Complete system, with or without casting base, includes internal cooling core, SuperSafe cover with attached leads, 4× glass plate pair sets, 4× combs (1,5More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Buffer volume, gel tank ~650 ml
Sample capacity 10 – 48
Unit W×H×D 250×140×200 mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR