Stopper, IN-Stopper
Supplier: B. Braun
4238010
720-2581EA 110 EUR
720-2581
Stopper, IN-Stopper
Stoppers PE/LDPE/HDPE Stoppers
IN-Stopper is a male luer lock closing cone with injection port for intermittent injections through injection membrane. It closes female accesses and offers a needle access via an injection membrane, e.g. to an infusion solution and has got a smooth, hygienic outer surface.
  • Smooth, hygienic outer surface
  • Offers a needle access via a membrane, which is not made with Latex
  • IN-Stopper can be used with alcohol and lipids
  • IN-Stopper are non-toxic and non-pyrogenic
  • Easy and convenient in handling
  • Five insertions possible
The device can be used for allMore Product Information