Integrated vertical electrophoresis unit, SE300 miniVE™
Supplier: HOEFER ELECTROPHORESIS
SE300 SE300-10A-1.0
732-1772EA 859 EUR
732-1772 732-1773
Integrated vertical electrophoresis unit, SE300 miniVE™
Electrophoresis Systems Vertical Gel Electrophoresis Systems
SE300 miniVE™ integrated vertical electrophoresis and blotting unit allows performance of both electrophoresis and electro transfer in one sturdy, compact unit.
  • Quick, excellent results using minimal buffer
  • Passive cooling produces gels with straight lanes and sharp bands
  • Transfer up to 4 gels in 45 minutes with only 300 ml buffer using optional SE302 blot modules
  • Designed for use with a wide variety of precast gels
Delivery information: Complete system includes lower buffer chamber (tank), gel modules with gaskets (2 assemblies), safety lid with highMore Product Information