Bioanalyzer High Sensitivity RNA Analysis
Supplier: AGILENT
5067-1513 5067-1514 5067-1535
AGLS5067-1513EA 1200 EUR
AGLS5067-1513 AGLS5067-1514 AGLS5067-1535
Bioanalyzer High Sensitivity RNA Analysis
Electrophoresis Systems Capillary Electrophoresis Systems
High sensitivity RNA sample quality assessment.
  • Maximum flexibility is enabled with options to check total RNA, mRNA, or cRNA quality
  • Minimal sample consumption required with only need 50 pg of total RNA for an effective analysis
  • Easily determine the RNA integrity number (RIN) using in-built software that automatically assigns an integrity number to a total RNA sample
  • This tool was designed to provide scientists an objective measurement of degradation in RNA samples
  • RNase free consumables avoid sample degradation during analysis
  • Accurate results are
More Product Information