RNA Kits
Supplier: AGILENT
DNF-471-0500 DNF-471-1000 DNF-472-0500 DNF-472-1000
AGLSDNF-471-0500EA 1440 EUR
AGLSDNF-471-0500 AGLSDNF-471-1000 AGLSDNF-472-0500 AGLSDNF-472-1000
RNA Kits
Electrophoresis Systems Capillary Electrophoresis Systems
Analysis of mRNA and total RNA quality control, IVT mRNA size, and detection of total RNA degradation.
  • Broad concentration range - The HS RNA kit (15 nt) has a concentration range of 50 to 5000 pg/µl, while the RNA kit (15 nt) covers 5 to 500 ng/µl
  • High and standard sensitivity kits - Minimise sample dilutions by choosing the kit that has the right concentration range for your needs
  • Reliable sizing - Sizing range from 200 to 6000 nt ensures accurate and precise sizing of RNA samples
  • Quality metrics - RNA Quality Number (RQN) quickly scores the integrity of
More Product Information