Rapamycin Streptomyces hygroscopicus

Supplier: Apollo Scientific
Warning

Synonyms: (3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,10,12,13,14,21,22,23,24,25,26,27,32,33,34,34a-Hexadecahydro-9,27-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-23,27-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine-1,5,11,28,29(4H,6H,31H)-pentone

BIR8101-250MG BIR8101-1G
APOSBIR8101-250MGEA 102 EUR
APOSBIR8101-250MG APOSBIR8101-1G
Rapamycin Streptomyces hygroscopicus
Rapamycin

Macrocyclic triene antibiotic.

Formula: C₅₁H₇₉NO₁₃
MW: 914.19 g/mol
Melting Pt: 183…185 °C
Storage Temperature: Freezer
MDL Number: MFCD00867594
CAS Number: 53123-88-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR