PGF2α high sensitivity ELISA kit

Supplier: ENZO LIFE SCIENCES

ADI930069 ADI931069
ENZOADI930069EA 517 EUR
ENZOADI930069 ENZOADI931069
PGF2α high sensitivity ELISA kit
Assays ELISAs

The PGF2α high sensitivity EIA kit for the quantitative determination of PGF2α in culture supernatants, milk, plasma, serum, saliva, and urine from any species.


  • Highly sensitive quantification of PGF2α
  • Assay more samples without extraction


The PGF2α high sensitivity EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 3 hours. Absorbance is read at 405 nm.


Ordering information: Kit contains DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.

Order Now

SPECIFICATIONS

Environmentally Preferable
Target protein PGF2α
Primary antibody reactivity
Assay Type Competitive
Host
Sensitivity 0,98 pg/ml (range 1,95 - 2000 pg/ml)
Storage conditions 4 °C (long term)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR