Anti-HMOX1 Mouse Monoclonal Antibody
Supplier: ENZO LIFE SCIENCES
ADIOSA111D ADIOSA111F ADIOSA111J
ENZOADIOSA111DEA 355 EUR
ENZOADIOSA111D ENZOADIOSA111F ENZOADIOSA111J
Anti-HMOX1 Mouse Monoclonal Antibody
Antibodies
Heme Oxygenase-1 (HO-1) also known as Hsp32, is the inducible isoform of heme oxygenase that catalyzes the NADPH, oxygen, and cytochrome P450 reductase dependent oxidation of heme to carbon monoxide, ferrous iron and biliverdin which is rapidly reduced to bilirubin. These products of the HO reaction have important physiological effects: carbon monoxide is a potent vasodilator and has been implicated to be a physiological regulator of cGMP and vascular tone; biliverdin and its product bilirubin are potent antioxidants; "free" iron increases oxidative stress and regulates the expression ofMore Product Information