Anti-IgG1 Mouse Monoclonal Antibody [clone: MOPC-21]

Supplier: ENZO LIFE SCIENCES
ADISAB600FI050 ADISAB600FI200
ENZOADISAB600FI050EA 290 EUR
ENZOADISAB600FI050 ENZOADISAB600FI200
Anti-IgG1 Mouse Monoclonal Antibody [clone: MOPC-21]
Antibodies
Type: Primary
Antigen: IgG1
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR