Mouse;Rat guanylin

Supplier: ENZO LIFE SCIENCES

ALX157011MC01
ENZOALX157011MC01EA 206 EUR
ENZOALX157011MC01
Mouse;Rat guanylin
Proteins and Peptides

Guanylyl cyclase activator


Endogenous intestinal guanylyl cyclase activator.

New Product

Order Now

SPECIFICATIONS

Protein/peptide name Guanylin
Environmentally Preferable
Species Rat,Mouse
Source
Purity >99%
Formulation Lyophilised

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR