Recombinant GroES (from E. coli)

Supplier: ENZO LIFE SCIENCES

ALX201142C025
ENZOALX201142C025EA 178 EUR
ENZOALX201142C025
Recombinant GroES (from E. coli)
Proteins and Peptides

Produced in E. coli.

New Product

Order Now

SPECIFICATIONS

Protein/peptide name GroES
Environmentally Preferable
Protein/peptide type Recombinant
Species
Source E. coli
Purity >99%
Concentration 4,2 mg/ml
Formulation Liquid. In 50 mM TRIS/HCl, pH 8,0, containing 10 mM KCl and 10 mM MgCl₂.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR