Human recombinant PARP1 (from Insect cells (Sf9)), HIS Tag

Supplier: ENZO LIFE SCIENCES

ALX201250C010
ENZOALX201250C010EA 444 EUR
ENZOALX201250C010
Human recombinant PARP1 (from Insect cells (Sf9)), HIS Tag
Proteins and Peptides

Produced in Sf21 cells. Human full-length PARP-1 is fused to a Myc-tag and a His-tag.

New Product

Order Now

SPECIFICATIONS

Protein/peptide name PARP1
Environmentally Preferable
Protein/peptide type Recombinant
Species Human
Source Insect cells (Sf9)
Purity >98% (SDS-PAGE)
Grade High purity
Concentration 0,5 mg/ml
Formulation Liquid. In 50 mM TRIS-HCl, pH 7,5, containing 100 mM sodium chloride, 50 mM imidazole, 0,2% NP-40 and 10% glycerol.
Tag sequence HIS Tag

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR