2-tert-Butylhydroquinone

Supplier: ENZO LIFE SCIENCES

ALX270061G010
ENZOALX270061G010EA 50.6 EUR
ENZOALX270061G010
2-tert-Butylhydroquinone
2-tert-Butylhydroquinone
Formula: C₁₀H₁₄O₂
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 128 °C
Density: 1,05 g/cm³ (20 °C)
Flash Pt: 171 °C
Storage Temperature: Ambient
MDL Number: MFCD00002344
CAS Number: 1948-33-0
EINECS: 217-752-2
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR