Cantharidin ≥98% (by TLC)

Supplier: ENZO LIFE SCIENCES

ALX270063M025 ALX270063M100
ENZOALX270063M025EA 103 EUR
ENZOALX270063M025 ENZOALX270063M100
Cantharidin ≥98% (by TLC)
Cantharidin

PP2A inhibitor


Potent protein phosphatase 2A (PP2A) inhibitor. At higher concentrations, also inhibits protein phosphatase 1 (PP1).

Formula: C₁₀H₁₂O₄
MDL Number: MFCD00134968
CAS Number: 56-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR