Vinyl-L-NIO ≥95% (by TLC)

Supplier: ENZO LIFE SCIENCES

Synonyms: L-VNIO, N5-(1-Imino-3-butenyl)-L-ornithine

ALX270216M025
ENZOALX270216M025EA 510 EUR
ENZOALX270216M025
Vinyl-L-NIO ≥95% (by TLC)
Vinyl-L-NIO

Very selective and potent neuronal nitric oxide synthase (nNOS/NOS I) inhibitor.


  • nNOS inhibitor


Clear film in the bottom of the vial.


Soluble in water, 100% ethanol (30mg/ml), DMSO (50mg/ml) or dimethyl formamide (50mg/ml).

Formula: C₉H₁₇N₃O₂·HCl
Storage Temperature: Refrigerator
CAS Number: 728944-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR