Sorafenib (BAY 43-9006) ≥95% (by HPLC)

Supplier: ENZO LIFE SCIENCES

Synonyms: 4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-2-pyridinecarboxamide, Sorafenib, N-[4-Chloro-3-(trifluoromethyl)phenyl]-N′-[4-[2-(N-methylcarbamoyl)-4-pyridyloxy]phenyl]urea, 4-(4-(3-(4-Chloro-3-(trifluoromethyl)phenyl)ureido)phenoxy)-N-methylpicolinamide

ALX270469M001 ALX270469M005 ALX270469M025
ENZOALX270469M001EA 101 EUR
ENZOALX270469M001 ENZOALX270469M005 ENZOALX270469M025
Sorafenib (BAY 43-9006) ≥95% (by HPLC)
Sorafenib (BAY 43-9006)

Broad-spectrum kinase inhibitor


Broad-spectrum kinase inhibitor.


Synthetic.

Formula: C₂₁H₁₆ClF₃N₄O₃
MW: 464.82 g/mol
MDL Number: MFCD06411450
CAS Number: 284461-73-0
New Product

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR