Kanamycin B sulphate ≥99% (by HPLC), high purity

Supplier: ENZO LIFE SCIENCES

ALX380095M250
ENZOALX380095M250EA 247 EUR
ENZOALX380095M250
Kanamycin B sulphate ≥99% (by HPLC), high purity
Kanamycin B sulphate

Antibiotic


Isolated from Streptomyces kanamyceticus.

Aminoglycoside antibiotic. Binds to the 70S ribosomal subunit. Inhibits translocation and elicits miscoding. Effective against Gram-negative and Gram-positive bacteria and mycoplasma.

Formula: C₁₈H₃₇N₅O₁₀·H₂SO₄
CAS Number: 29701-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR