DICA ≥98% (by TLC)

Supplier: ENZO LIFE SCIENCES

Synonyms: 2-(2,4-Dichlorophenoxy)-N-(2-mercaptoethyl)acetamide

ALX420036M025
ENZOALX420036M025EA 306 EUR
ENZOALX420036M025
DICA ≥98% (by TLC)
DICA

Inhibitor of caspase-3 and caspase-7 binding to an allosteric site and not to the active site.


  • Inhibitor of caspase-3 and -7


White to off-white solid.


Soluble in methanol or DMSO.

Formula: C₁₀H₁₁Cl₂NO₂S
MW: 280.2 g/mol
Storage Temperature: Freezer
CAS Number: 613665-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR