Copper(II) protoporphyrin IX Free_acid

Supplier: ENZO LIFE SCIENCES

ALX430048M025
ENZOALX430048M025EA 153 EUR
ENZOALX430048M025
Copper(II) protoporphyrin IX Free_acid
Copper(II) protoporphyrin IX

Negative control for the heme oxygenase inhibitor zinc protoporphyrin IX (Prod. No. ALX-430-049).

Formula: C₃₄H₃₂CuN₄O₄
MW: 624.2 g/mol
MDL Number: MFCD01076447
CAS Number: 14494-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR