Ionomycin calcium salt

Supplier: ENZO LIFE SCIENCES

ALX450007M001 ALX450007M005
ENZOALX450007M001EA 165 EUR
ENZOALX450007M001 ENZOALX450007M005
Ionomycin calcium salt
Ionomycin calcium salt
Formula: C₄₁H₇₀CaO₉
MDL Number: MFCD00151183
CAS Number: 56092-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR