Folic acid

Supplier: ENZO LIFE SCIENCES

Synonyms: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9

ALX460006G010
ENZOALX460006G010EA 52.8 EUR
ENZOALX460006G010
Folic acid
Folic acid
Formula: C₁₉H₁₉N₇O₆
MDL Number: MFCD00079305
CAS Number: 59-30-3
EINECS: 200-419-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR